4-(Chloromethyl)phenyltrichlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

L16431-06 L16431-14 L16431-22
CAAAAL16431-06EA 75.21 CAD
CAAAAL16431-06 CAAAAL16431-14 CAAAAL16431-22
4-(Chloromethyl)phenyltrichlorosilane 97%
4-(Chloromethyl)phenyltrichlorosilane

Purity: 97% CAS No.: 13688-90-9 Molecular Formula: C7H6Cl4Si Formula Weight: 260.02 Flash Point: 78°(172°F) Refractive Index: 1.5480 Boiling Point: 142-144°/15mm Beilstein Reference: 3090295 UN No.: 2987 MDL No.: MFCD00045268

Formula: C₇H₆Cl₄Si
MW: 260.02 g/mol
Boiling Pt: 142…144 °C/15 mm
Density: 1.361
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045268
CAS Number: 13688-90-9
EINECS: 237-203-0
UN: 2987
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR