Ethyl pyrrole-2-carboxylate 98+%
Supplier: Thermo Scientific Chemicals
L16382-06
L16382-14
CAAAAL16382-06EA
133.35
CAD
CAAAAL16382-06
CAAAAL16382-14
Ethyl pyrrole-2-carboxylate 98+%
Ethyl pyrrole-2-carboxylate
Purity: 98+%
CAS No.: 2199-43-1
Molecular Formula: C7H9NO2
Formula Weight: 139.16
Melting Point: 37-39°
Boiling Point: 70°/0.8mm
MDL No.: MFCD00817049
Formula:
C₇H₉NO₂ Boiling Pt: 70 °C/0.8 mm Melting Pt: 37…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00817049 CAS Number: 2199-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...