Ethyl-3-methylisoxazole-4-carboxylate 97%
Supplier: Thermo Scientific Chemicals
L16362-03
L16362-06
CAAAAL16362-03EA
62.41
CAD
CAAAAL16362-03
CAAAAL16362-06
Ethyl-3-methylisoxazole-4-carboxylate 97%
Ethyl-3-methylisoxazole-4-carboxylate
Purity: 97%
CAS No.: 20328-15-8
Molecular Formula: C7H9NO3
Formula Weight: 155.15
Refractive Index: 1.4595
Boiling Point: 64°/0.6mm
Beilstein Reference: 1072756
MDL No.: MFCD01075736
Formula:
C₇H₉NO₃ Boiling Pt: 64 °C/0.6 mm Storage Temperature: Ambient |
MDL Number:
MFCD01075736 CAS Number: 20328-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...