(S)-(-)-1-(p-Tolyl)ethylamine ≥98%, ee 99%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16329-03
L16329-06
L16329-14
CAAAAL16329-03EA
76
CAD
CAAAAL16329-03
CAAAAL16329-06
CAAAAL16329-14
(S)-(-)-1-(p-Tolyl)ethylamine ≥98%, ee 99%, ChiPros®
(S)-(-)-1-(p-Tolyl)ethylamine
Purity: 98%
CAS No.: 27298-98-2
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 82°(179°F)
Refractive Index: 1.5215
Melting Point:
Formula:
C₉H₁₃N Boiling Pt: 204 °C Melting Pt: < –20 °C Density: 0.94 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00145246 CAS Number: 27298-98-2 EINECS: 623-635-0 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...