Trimorpholinophosphine oxide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phosphoric Trimorpholide
L16315-06
L16315-14
CAAAAL16315-06EA
69.36
CAD
CAAAAL16315-06
CAAAAL16315-14
Trimorpholinophosphine oxide 99%
Trimorpholinophosphine oxide
Purity: 99%
CAS No.: 4441-12-7
Molecular Formula: C12H24N3O4P
Formula Weight: 305.32
Melting Point: 188-192°
Beilstein Reference: 280155
MDL No.: MFCD00047406
Formula:
C₁₂H₂₄N₃O₄P MW: 305.31 g/mol Melting Pt: 188…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047406 CAS Number: 4441-12-7 EINECS: 224-662-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...