N-(4-Chloro-2-butynyl)phthalimide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16313-03
L16313-06
CAAAAL16313-03EA
135.25
CAD
CAAAAL16313-03
CAAAAL16313-06
N-(4-Chloro-2-butynyl)phthalimide 97%
N-(4-Chloro-2-butynyl)phthalimide
Purity: 97%
CAS No.: 4819-69-6
Molecular Formula: C12H8ClNO2
Formula Weight: 233.66
Odor: Nearly odorless
Melting Point: 115-120°
Beilstein Reference: 179626
UN No.: 3261
MDL No.: MFCD01318120
Formula:
C₁₂H₈ClNO₂ Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318120 CAS Number: 4819-69-6 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...