2-Carboxyphenylboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16301-03
L16301-06
L16301-MD
CAAAAL16301-03EA
377.62
CAD
CAAAAL16301-03
CAAAAL16301-06
CAAAAL16301-MD
2-Carboxyphenylboronic acid 95%
2-Carboxyphenylboronic acid
Purity: 95%
CAS No.: 149105-19-1
Molecular Formula: C7H7BO4
Formula Weight: 165.94
Melting Point: ca 220°
Beilstein Reference: 2835407
Formula:
C₇H₇BO₄ Melting Pt: ca 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318118 CAS Number: 149105-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...