4-Amino-2-isopropyl-5-methylphenol hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

L16282-03 L16282-06
CAAAAL16282-03EA 37.13 CAD
CAAAAL16282-03 CAAAAL16282-06
4-Amino-2-isopropyl-5-methylphenol hydrochloride 97%
4-Amino-2-isopropyl-5-methylphenol hydrochloride

Purity: 97% CAS No.: 6321-11-5 Molecular Formula: C10H15NO.HCl Formula Weight: 201.70 Odor: Odorless Melting Point: 258-262° Beilstein Reference: 3700651 MDL No.: MFCD00035504

Formula: C₁₀H₁₆ClNO
MW: 201.7 g/mol
Melting Pt: 258…262 °C
Storage Temperature: Ambient
MDL Number: MFCD00035504
CAS Number: 6321-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR