N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol), Ethylenediamine-N,N,N',N'-tetra-2-propanol

L16280-AE L16280-AP
CAAAAL16280-AEEA 56.41 CAD
CAAAAL16280-AE CAAAAL16280-AP
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 99%
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine

Purity: 99% CAS No.: 102-60-3 Molecular Formula: C14H32N2O4 Formula Weight: 292.42 Flash Point: 110°(230°F) Refractive Index: 1.4800 Odor: Odorless Boiling Point: 240°/10mm Merck Reference: 14,3599 MDL No.: MFCD00004534

Formula: C₁₄H₃₂N₂O₄
MW: 292.42 g/mol
Boiling Pt: 240 °C/10 mm
Density: 1.030
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004534
CAS Number: 102-60-3
EINECS: 203-041-4
Merck Index: 14,3599

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR