1,5-Pentamethylene-1H-tetrazole 98%

Supplier: Thermo Scientific Chemicals
Danger

L16185-06 L16185-14
CAAAAL16185-06EA 54.83 CAD
CAAAAL16185-06 CAAAAL16185-14
1,5-Pentamethylene-1H-tetrazole 98%
1,5-Pentamethylene-1H-tetrazole

Purity: 98% CAS No.: 54-95-5 Molecular Formula: C6H10N4 Formula Weight: 138.17 Melting Point: 59-61° Boiling Point: 194°/12mm Merck Reference: 14,7141 UN No.: 2811 MDL No.: MFCD00005939

Formula: C₆H₁₀N₄
Boiling Pt: 194 °C/12 mm
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00005939
CAS Number: 54-95-5
EINECS: 200-219-3
UN: 2811
ADR: "6.1",III
Merck Index: 14,7141

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR