4-Bromo-3-methylbenzamide 99%

Supplier: Thermo Scientific Chemicals

L16176-03 L16176-06 L16176-14
CAAAAL16176-03EA 22.28 CAD
CAAAAL16176-03 CAAAAL16176-06 CAAAAL16176-14
4-Bromo-3-methylbenzamide 99%
4-Bromo-3-methylbenzamide

Purity: 99% CAS No.: 170229-98-8 Molecular Formula: C8H8BrNO Formula Weight: 214.06 Melting Point: 177-182° Beilstein Reference: 868074

Formula: C₈H₈BrNO
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00672914
CAS Number: 170229-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR