4,4'-(Hexafluoroisopropylidene)diphthalic anhydride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane

L16150-06 L16150-14 L16150-22
CAAAAL16150-06EA 107.44 CAD
CAAAAL16150-06 CAAAAL16150-14 CAAAAL16150-22
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride 99%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride

Purity: 99% CAS No.: 1107-00-2 Molecular Formula: C19H6F6O6 Formula Weight: 444.25 Melting Point: 244-247° Beilstein Reference: 7057916

Formula: C₈H₄O₃
MW: 444.25 g/mol
Melting Pt: 244…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00039143
CAS Number: 1107-00-2
EINECS: 214-170-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR