Amberlite® XAD-1180, Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

L16097-22 L16097-36
CAAAAL16097-22EA 50.72 CAD
CAAAAL16097-22 CAAAAL16097-36
Amberlite® XAD-1180, Amberlite™ ion exchange resin
Amberlite® XAD-1180

CAS No.: 97396-56-0 MDL No.: MFCD00145830

Storage Temperature: Ambient
MDL Number: MFCD00145830
CAS Number: 97396-56-0
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR