6-Bromo-2-naphthoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16093-06
L16093-14
CAAAAL16093-06EA
83.9
CAD
CAAAAL16093-06
CAAAAL16093-14
6-Bromo-2-naphthoic acid 98%
6-Bromo-2-naphthoic acid
Purity: 98%
CAS No.: 5773-80-8
Molecular Formula: C11H7BrO2
Formula Weight: 251.08
Melting Point: 292-296°
Beilstein Reference: 2328940
Formula:
C₁₁H₇BrO₂ Melting Pt: 292…296 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075720 CAS Number: 5773-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...