Bis(pinacolato)diboron 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

L16088-03 L16088-06 L16088-14
CAAAAL16088-03EA 65.1 CAD
CAAAAL16088-03 CAAAAL16088-06 CAAAAL16088-14
Bis(pinacolato)diboron 98+%
Bis(pinacolato)diboron

Purity: 98+% CAS No.: 73183-34-3 Molecular Formula: C12H24B2O4 Formula Weight: 253.94 Storage Temperature: 2°C to 8°C Melting Point: 133-139° Beilstein Reference: 7703552 MDL No.: MFCD00799570

Formula: C₁₂H₂₄B₂O₄
MW: 253.94 g/mol
Melting Pt: 133…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR