4-[(Trifluoromethyl)thio]aniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-((Trifluoromethyl)thio)aniline
L16067-03
L16067-06
L16067-14
CAAAAL16067-03EA
42.19
CAD
CAAAAL16067-03
CAAAAL16067-06
CAAAAL16067-14
4-[(Trifluoromethyl)thio]aniline 98%
4-[(Trifluoromethyl)thio]aniline
Purity: 98%
CAS No.: 372-16-7
Molecular Formula: C7H6F3NS
Formula Weight: 193.19
Flash Point: 106°(222°F)
Refractive Index: 1.5280
Odor: Unpleasant
Boiling Point: 103°/11mm
Beilstein Reference: 2208859
UN No.: 2810
MDL No.: MFCD00040926
Formula:
C₇H₆F₃NS MW: 193.19 g/mol Boiling Pt: 103 °C/11 mm Density: 1.344 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040926 CAS Number: 372-16-7 EINECS: 206-744-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...