Diethyl difluoromalonate 97%

Supplier: Thermo Scientific Chemicals
Warning

L16042-03 L16042-06
CAAAAL16042-03EA 116.45 CAD
CAAAAL16042-03 CAAAAL16042-06
Diethyl difluoromalonate 97%
Diethyl difluoromalonate

Purity: 97% CAS No.: 680-65-9 Molecular Formula: C7H10F2O4 Formula Weight: 196.15 Refractive Index: 1.3800 Boiling Point: 94-95°/23mm MDL No.: MFCD00221617

Formula: C₇H₁₀F₂O₄
Boiling Pt: 94…95 °C/23 mm
Density: 1.179
Storage Temperature: Ambient
MDL Number: MFCD00221617
CAS Number: 680-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR