2-Isocyanato-1-methoxy-4-methylbenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16001-03
L16001-06
CAAAAL16001-03EA
40.13
CAD
CAAAAL16001-03
CAAAAL16001-06
2-Isocyanato-1-methoxy-4-methylbenzene 97%
2-Isocyanato-1-methoxy-4-methylbenzene
Purity: 97%
CAS No.: 59741-04-7
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Flash Point:
Formula:
C₉H₉NO₂ Boiling Pt: 240 °C Density: 1.112 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013869 CAS Number: 59741-04-7 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...