trans-1,4-Cyclohexane diisocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15991-03
L15991-06
CAAAAL15991-03EA
82.16
CAD
CAAAAL15991-03
CAAAAL15991-06
trans-1,4-Cyclohexane diisocyanate 97%
trans-1,4-Cyclohexane diisocyanate
Purity: 97%
CAS No.: 7517-76-2
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Odor: Irritating
Melting Point: 59-62°
Boiling Point: 145-146°/32mm
Beilstein Reference: 2804862
UN No.: 2811
MDL No.: MFCD00074787
Formula:
C₈H₁₀N₂O₂ Boiling Pt: 145…146 °C/32 mm Melting Pt: 59…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074787 CAS Number: 7517-76-2 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...