4-Acetamidobenzenesulfonyl azide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15916-06
L15916-14
CAAAAL15916-06EA
51.67
CAD
CAAAAL15916-06
CAAAAL15916-14
4-Acetamidobenzenesulfonyl azide 97%
4-Acetamidobenzenesulphonyl azide
Fieser: 14,1
Purity: 97%
CAS No.: 2158-14-7
Molecular Formula: C8H8N4O3S
Formula Weight: 240.24
Storage Temperature: -30°C to -10°C
Melting Point: 107-111°
Beilstein Reference: 2219568
MDL No.: MFCD00029626
Formula:
C₈H₈N₄O₃S MW: 240.24 g/mol Melting Pt: 107…111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00029626 CAS Number: 2158-14-7 EINECS: 606-801-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...