1-(2-Pyrimidyl)piperazine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(1-Piperazinyl)pyrimidine, 2-(Piperazin-1-yl)pyrimidine
L15884-06
L15884-14
L15884-22
CAAAAL15884-06EA
51.82
CAD
CAAAAL15884-06
CAAAAL15884-14
CAAAAL15884-22
1-(2-Pyrimidyl)piperazine 99%
1-(2-Pyrimidyl)piperazine
Purity: 99%
CAS No.: 20980-22-7
Molecular Formula: C8H12N4
Formula Weight: 164.21
Melting Point: 31-35°
Boiling Point: 118-120°/2mm
Beilstein Reference: 151178
UN No.: 3259
MDL No.: MFCD00040742
Formula:
C₈H₁₂N₄ Boiling Pt: 118…120 °C/2 mm Melting Pt: 31…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040742 CAS Number: 20980-22-7 EINECS: 244-135-5 UN: 3259 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...