2-(4-Aminophenyl)ethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(2-Aminoethyl)aniline, 1-Amino-2-(4-aminophenyl)ethane, 4-Aminophenethylamine
L15782-03
L15782-06
CAAAAL15782-03EA
95.12
CAD
CAAAAL15782-03
CAAAAL15782-06
2-(4-Aminophenyl)ethylamine 98%
2-(4-Aminophenyl)ethylamine
Purity: 98%
CAS No.: 13472-00-9
Molecular Formula: C8H12N2
Formula Weight: 136.20
Flash Point:
Formula:
C₈H₁₂N₂ Boiling Pt: 103 °C/0.3 mm Melting Pt: 28…31 °C Density: 1.057 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008194 CAS Number: 13472-00-9 EINECS: 236-739-2 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...