Dess-Martin periodinane

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

L15779-03 L15779-06
CAAAAL15779-03EA 62.73 CAD
CAAAAL15779-03 CAAAAL15779-06
Dess-Martin periodinane
Dess-Martin periodinane

Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.


CAS No.: 87413-09-0 Molecular Formula: C13H13IO8 Formula Weight: 424.15 Storage Temperature: -30°C to -10°C Melting Point: 130-132° dec. Beilstein Reference: 4548207 UN No.: 1479 MDL No.: MFCD00130127

Formula: C₁₃H₁₃IO₈
Melting Pt: 130…132 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1479
ADR: "5.1",III
Merck Index: 14,2933

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR