Methylenediphenyl-4,4'-diisocyanate 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4,4'-Diphenylmethane Diisocyanate, Methylene di-p-phenyl diisocyanate, Diphenylmethane 4,4'-diisocyanate, Bis(4-isocyanatophenyl)methane, MDI, 4,4'-Methylenebis(phenyl Isocyanate)
L15740-14
L15740-22
L15740-36
CAAAAL15740-14EA
47.08
CAD
CAAAAL15740-14
CAAAAL15740-22
CAAAAL15740-36
Methylenediphenyl-4,4'-diisocyanate 98% stabilised
Methylenediphenyl-4,4'-diisocyanate
Purity: 98%
CAS No.: 101-68-8
Molecular Formula: C15H10N2O2
Formula Weight: 250.26
Storage Temperature: -30°C to -10°C
Flash Point: 196°(385°F)
Odor: Weak, characteristic
Melting Point: 41-44°
Boiling Point: 194°/5mm
Beilstein Reference: 797662
UN No.: 2811
MDL No.: MFCD00036131
Formula:
C₁₅H₁₀N₂O₂ MW: 250.26 g/mol Boiling Pt: 194 °C/5 mm Melting Pt: 41…44 °C Density: 1.239 Flash Pt: 196 °C(385 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00036131 CAS Number: 101-68-8 EINECS: 202-966-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...