[Hydroxy(tosyloxy)iodo]benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15701-06
L15701-14
CAAAAL15701-06EA
66.68
CAD
CAAAAL15701-06
CAAAAL15701-14
[Hydroxy(tosyloxy)iodo]benzene 97%
[Hydroxy(tosyloxy)iodo]benzene
Fieser: 14,179 16,179 17,150 18,187 19,170 20,193 21,223
Purity: 97%
CAS No.: 27126-76-7
Molecular Formula: C13H13IO4S
Formula Weight: 392.22
Melting Point: 131-136°
Beilstein Reference: 2150074
MDL No.: MFCD00011547
Formula:
C₁₃H₁₃IO₄S Melting Pt: 131…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011547 CAS Number: 27126-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...