4-Chloro-3-nitro-2-pyridone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15686-06
L15686-14
CAAAAL15686-06EA
145.68
CAD
CAAAAL15686-06
CAAAAL15686-14
4-Chloro-3-nitro-2-pyridone 98%
4-Chloro-3-nitro-2-pyridone
Purity: 98%
CAS No.: 165547-79-5
Molecular Formula: C5H3ClN2O3
Formula Weight: 174.54
Melting Point: 222-224°
Beilstein Reference: 149557
MDL No.: MFCD03093067
Formula:
C₅H₃ClN₂O₃ Melting Pt: 222…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093067 CAS Number: 165547-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...