4,4'-Oxybis(benzoic acid) ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4′-Oxybis(benzoic acid)
L15651-14
L15651-22
CAAAAL15651-14EA
61.15
CAD
CAAAAL15651-14
CAAAAL15651-22
4,4'-Oxybis(benzoic acid) ≥98%
4,4'-Oxybis(benzoic acid)
Purity: 98+%
CAS No.: 2215-89-6
Molecular Formula: C14H10O5
Formula Weight: 258.23
Melting Point: 329-332°
Beilstein Reference: 2218315
MDL No.: MFCD00013988
Formula:
C₁₄H₁₀O₅ MW: 258.23 g/mol Melting Pt: 329…332 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013988 CAS Number: 2215-89-6 EINECS: 218-683-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...