L-Proline benzyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Pro-OBzl.HCl
L15618-03
L15618-06
CAAAAL15618-03EA
36.34
CAD
CAAAAL15618-03
CAAAAL15618-06
L-Proline benzyl ester hydrochloride 98%
L-Proline benzyl ester hydrochloride
Purity: 98%
CAS No.: 16652-71-4
Molecular Formula: C12H15NO2.HCl
Formula Weight: 241.72
Melting Point: 146-148°
Beilstein Reference: 3598081
Optical Rotation: -44° (c=1 in methanol)
MDL No.: MFCD00039002
Formula:
C₁₂H₁₅NO₂ MW: 241.72 g/mol Melting Pt: 146…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039002 CAS Number: 16652-71-4 EINECS: 240-700-5 |
Specification Test Results
Optical rotation | -44° (c=1 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...