2,2-Diphenylpropylamine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
L15615-03
L15615-06
CAAAAL15615-03EA
76.95
CAD
CAAAAL15615-03
CAAAAL15615-06
2,2-Diphenylpropylamine hydrochloride 98+%
2,2-Diphenylpropylamine hydrochloride
Purity: 98+%
CAS No.: 40691-66-5
Molecular Formula: C15H17N.HCl
Formula Weight: 247.77
Melting Point: 250-256°
Beilstein Reference: 3709987
Formula:
C₁₅H₁₈ClN Melting Pt: 250…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008133 CAS Number: 40691-66-5 EINECS: 255-039-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...