2,3-Dihydro-1H-benz[de]isoquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15613-03
L15613-MD
CAAAAL15613-03EA
263.86
CAD
CAAAAL15613-03
CAAAAL15613-MD
2,3-Dihydro-1H-benz[de]isoquinoline 97%
2,3-Dihydro-1H-benz[de]isoquinoline
Purity: 97%
CAS No.: 22817-26-1
Molecular Formula: C12H11N
Formula Weight: 169.23
Melting Point: 96-101°
Boiling Point: 120°/0.5mm
Beilstein Reference: 134303
UN No.: 3259
MDL No.: MFCD01075706
Formula:
C₁₂H₁₁N Boiling Pt: 120 °C/0.5 mm Melting Pt: 96…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075706 CAS Number: 22817-26-1 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...