2,3-Diphenylmaleic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15612-03
L15612-06
CAAAAL15612-03EA
79
CAD
CAAAAL15612-03
CAAAAL15612-06
2,3-Diphenylmaleic anhydride 98%
2,3-Diphenylmaleic anhydride
Purity: 98%
CAS No.: 4808-48-4
Molecular Formula: C16H10O3
Formula Weight: 250.26
Melting Point: 157-160°
Boiling Point: 236°/15mm
MDL No.: MFCD00005521
Formula:
C₁₆H₁₀O₃ Boiling Pt: 236 °C/15 mm Melting Pt: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005521 CAS Number: 4808-48-4 EINECS: 225-370-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...