2-Iodosobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15606-03
L15606-06
CAAAAL15606-03EA
46.77
CAD
CAAAAL15606-03
CAAAAL15606-06
2-Iodosobenzoic acid 97%
2-Iodosobenzoic acid
Commonly used peptide reagent for cleavage at tryptophyl and tyrosyl residues
Purity: 97%
CAS No.: 304-91-6
Molecular Formula: C7H5IO3
Formula Weight: 264.02
Melting Point: ca 230° dec.
Beilstein Reference: 1939973
Formula:
C₇H₅IO₃ Melting Pt: ca 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002401 CAS Number: 304-91-6 EINECS: 206-159-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...