Cycloheptylacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15553-03
L15553-06
CAAAAL15553-03EA
105.54
CAD
CAAAAL15553-03
CAAAAL15553-06
Cycloheptylacetic acid 99%
Cycloheptylacetic acid
Purity: 99%
CAS No.: 4401-20-1
Molecular Formula: C9H16O2
Formula Weight: 156.23
Refractive Index: 1.4725
Melting Point: 14-15°
Boiling Point: 110-112°/1mm
Beilstein Reference: 1935484
MDL No.: MFCD01075701
Formula:
C₉H₁₆O₂ Boiling Pt: 110…112 °C/1 mm Melting Pt: 14…15 °C Density: 1.023 Storage Temperature: Ambient |
MDL Number:
MFCD01075701 CAS Number: 4401-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...