Benzyl-tert-butyl malonate 95%
Supplier: Thermo Scientific Chemicals
L15533-06
L15533-14
CAAAAL15533-06EA
125.61
CAD
CAAAAL15533-06
CAAAAL15533-14
Benzyl-tert-butyl malonate 95%
Benzyl-tert-butyl malonate
Purity: 95%
CAS No.: 72594-86-6
Molecular Formula: C14H18O4
Formula Weight: 250.30
Refractive Index: 1.4860
Beilstein Reference: 5434466
MDL No.: MFCD01075175
Formula:
C₁₄H₁₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD01075175 CAS Number: 72594-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...