3-Isopropylphenylboronic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L15530-03 L15530-MD
CAAAAL15530-03EA 197.5 CAD
CAAAAL15530-03 CAAAAL15530-06 CAAAAL15530-MD
3-Isopropylphenylboronic acid 99%
3-Isopropylphenylboronic acid

Purity: 99% CAS No.: 216019-28-2 Molecular Formula: C9H13BO2 Formula Weight: 164.01 Flash Point: 110°(230°F) Melting Point: ca 75-80° Beilstein Reference: 3090001 MDL No.: MFCD01074634

Formula: C₉H₁₃BO₂
Melting Pt: ca 75…80 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01074634
CAS Number: 216019-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR