2,3-Dibromofuran 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with calcium carbonate 0.5%

L15518-03 L15518-06
CAAAAL15518-03EA 80.9 CAD
CAAAAL15518-03 CAAAAL15518-06
2,3-Dibromofuran 97% stabilized
2,3-Dibromofuran

Stab. with 0.5% calcium carbonate


Purity: 97% CAS No.: 30544-34-4 Molecular Formula: C4H2Br2O Formula Weight: 225.87 Storage Temperature: -30°C to -10°C Refractive Index: 1.5470 Boiling Point: 75-78°/75mm Beilstein Reference: 107511 MDL No.: MFCD01074839

Formula: C₄H₂Br₂O
Boiling Pt: 75…78 °C/75 mm
Density: 2.000
Storage Temperature: Refrigerator
MDL Number: MFCD01074839
CAS Number: 30544-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR