4-(Benzyloxy)-2-hydroxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

L15515-03 L15515-06
CAAAAL15515-03EA 58.93 CAD
CAAAAL15515-03 CAAAAL15515-06
4-(Benzyloxy)-2-hydroxybenzaldehyde 99%
4-(Benzyloxy)-2-hydroxybenzaldehyde

Purity: 99% CAS No.: 52085-14-0 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 77-80° Beilstein Reference: 982995

Formula: C₁₄H₁₂O₃
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD01075698
CAS Number: 52085-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR