2-Bromo-4'-(1-pyrrolidinyl)acetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15407-06
L15407-14
CAAAAL15407-06EA
138.09
CAD
CAAAAL15407-06
CAAAAL15407-14
2-Bromo-4'-(1-pyrrolidinyl)acetophenone 97%
2-Bromo-4'-(1-pyrrolidinyl)acetophenone
Purity: 97%
CAS No.: 216144-18-2
Molecular Formula: C12H14BrNO
Formula Weight: 268.16
Melting Point: 120-122°
Beilstein Reference: 8549816
UN No.: 3261
MDL No.: MFCD01075690
Formula:
C₁₂H₁₄BrNO Melting Pt: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075690 CAS Number: 216144-18-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...