L-Pipecolic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline

L15373-03 L15373-06
CAAAAL15373-03EA 199.08 CAD
CAAAAL15373-03 CAAAAL15373-06
L-Pipecolic acid 99%
L-Pipecolic acid

Purity: 99% CAS No.: 3105-95-1 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Melting Point: ca 270° dec. Beilstein Reference: 81093 Optical Rotation: -26° (c=1 in water) MDL No.: MFCD00005981

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: ca 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005981
CAS Number: 3105-95-1
EINECS: 221-462-1
Merck Index: 14,7458

Order Now

Specification Test Results

Optical rotation -26° (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR