Bzl-Gly-OH ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Benzylamino)acetic acid, N-Benzylglycine

L15368-06 L15368-14
CAAAAL15368-06EA 96.22 CAD
CAAAAL15368-06 CAAAAL15368-14
Bzl-Gly-OH ≥98%
Bzl-Gly-OH

Purity: 98+% CAS No.: 17136-36-6 Molecular Formula: C6H5CH2NHCH2CO2H Formula Weight: 165.19 Melting Point: ca 210° dec. Beilstein Reference: 2209788 MDL No.: MFCD00190772

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: ∼ 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00190772
CAS Number: 17136-36-6
EINECS: 241-194-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR