Sodium vinylsulfonate ∼25% in aqueous solution, Technical Grade

Supplier: Thermo Scientific Chemicals

L15356-AE L15356-AP L15356-AU
CAAAAL15356-AEEA 56.88 CAD
CAAAAL15356-AE CAAAAL15356-AP CAAAAL15356-AU
Sodium vinylsulfonate ∼25% in aqueous solution, Technical Grade
Sodium vinylsulfonate

CAS No.: 3039-83-6 Molecular Formula: CH2=CHSO3Na Formula Weight: 130.10 Refractive Index: 1.3770 MDL No.: MFCD00007520

Formula: C₂H₄O₃S
Density: 1.180
Storage Temperature: Ambient
MDL Number: MFCD00007520
CAS Number: 3039-83-6
EINECS: 221-242-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR