Pyrrole-2-carbonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

L15330-03 L15330-06
CAAAAL15330-03EA 91.32 CAD
CAAAAL15330-03 CAAAAL15330-06
Pyrrole-2-carbonitrile 99%
Pyrrole-2-carbonitrile

Purity: 99% CAS No.: 4513-94-4 Molecular Formula: C5H4N2 Formula Weight: 92.10 Refractive Index: 1.5500 Boiling Point: 92-94°/2mm Beilstein Reference: 107047 UN No.: 3276 MDL No.: MFCD00234061

Formula: C₅H₄N₂
Boiling Pt: 92…94 °C/2 mm
Density: 1.086
Storage Temperature: Ambient
MDL Number: MFCD00234061
CAS Number: 4513-94-4
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR