Methyl isopropyl carbonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15314-03
L15314-06
L15314-14
CAAAAL15314-03EA
57.04
CAD
CAAAAL15314-03
CAAAAL15314-06
CAAAAL15314-14
Methyl isopropyl carbonate 97%
Methyl isopropyl carbonate
Purity: 97%
CAS No.: 51729-83-0
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 27°(80°F)
Refractive Index: 1.3830
Boiling Point: 116-118°
Beilstein Reference: 1744735
UN No.: 3272
MDL No.: MFCD01075689
Formula:
C₅H₁₀O₃ Boiling Pt: 116…118 °C Density: 0.975 Flash Pt: 27 °C(80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01075689 CAS Number: 51729-83-0 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...