4,4-Dimethyl-1-phenylpentane-1,3-dione 99%

Supplier: Thermo Scientific Chemicals

L15251-06 L15251-14
CAAAAL15251-06EA 108.23 CAD
CAAAAL15251-06 CAAAAL15251-14
4,4-Dimethyl-1-phenylpentane-1,3-dione 99%
4,4-Dimethyl-1-phenylpentane-1,3-dione

Purity: 99% CAS No.: 13988-67-5 Molecular Formula: C13H16O2 Formula Weight: 204.27 Refractive Index: 1.5590 Boiling Point: 127-128°/2mm Beilstein Reference: 1868436 MDL No.: MFCD00511275

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Boiling Pt: 127…128 °C/2 mm
Density: 1.024
Storage Temperature: Ambient
MDL Number: MFCD00511275
CAS Number: 13988-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR