3-Aminophthalhydrazide monosodium salt ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L15205-03 L15205-06
CAAAAL15205-03EA 94.17 CAD
CAAAAL15205-03 CAAAAL15205-06
3-Aminophthalhydrazide monosodium salt ≥98%
3-Aminophthalhydrazide monosodium salt

Purity: 98+% CAS No.: 20666-12-0 Molecular Formula: C8H6N3NaO2 Formula Weight: 199.15 MDL No.: MFCD00161491

Formula: C₈H₆N₃NaO₂
Storage Temperature: Ambient
MDL Number: MFCD00161491
CAS Number: 20666-12-0
EINECS: 629-344-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR