N-Benzylglycine hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15200-14
L15200-22
CAAAAL15200-06EA
94.17
CAD
CAAAAL15200-06
CAAAAL15200-14
CAAAAL15200-22
N-Benzylglycine hydrochloride ≥98%
N-Benzylglycine hydrochloride
Purity: 98+%
CAS No.: 7689-50-1
Molecular Formula: C6H5CH2NHCH2CO2H.HCl
Formula Weight: 201.65
Melting Point: ca 220° dec.
Beilstein Reference: 3915652
MDL No.: MFCD00156934
Formula:
C₉H₁₁NO₂·HCl Melting Pt: ca 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00156934 CAS Number: 7689-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...