Amberlite® IR-120 (Na), Amberlite™ ion exchange resin
Supplier: Thermo Scientific Chemicals
L15199-0E
L15199-22
L15199-36
CAAAAL15199-0EEA
268.6
CAD
CAAAAL15199-0E
CAAAAL15199-22
CAAAAL15199-36
Amberlite® IR-120 (Na), Amberlite™ ion exchange resin
Amberlite® IR-120 (Na)
Water softening, dealkalinization, deionization
Fieser: 1,511 4,266 5,355 6,302 9,256
CAS No.: 78922-04-0
MDL No.: MFCD00132707
Storage Temperature:
Ambient |
MDL Number:
MFCD00132707 CAS Number: 9002-23-7 Merck Index: 14,382 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...