4-(Hydroxymethyl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15194-03
L15194-06
CAAAAL15194-03EA
65.41
CAD
CAAAAL15194-03
CAAAAL15194-06
4-(Hydroxymethyl)phenylboronic acid 98%
4-(Hydroxymethyl)phenylboronic acid
Purity: 98%
CAS No.: 59016-93-2
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: ca 280°
Beilstein Reference: 2831413
Formula:
C₇H₉BO₃ Melting Pt: ca 280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792672 CAS Number: 59016-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...