Diethylenetriaminepentaacetic dianhydride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15178-03
L15178-06
CAAAAL15178-03EA
82
CAD
CAAAAL15178-03
CAAAAL15178-06
Diethylenetriaminepentaacetic dianhydride 95%
Diethylenetriaminepentaacetic dianhydride
Purity: 95%
CAS No.: 23911-26-4
Molecular Formula: C14H19N3O8
Formula Weight: 357.32
Melting Point: ca 190° dec.
Beilstein Reference: 1230954
MDL No.: MFCD00010697
Formula:
C₁₄H₁₉N₃O₈ Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010697 CAS Number: 23911-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...