(tert-Butoxycarbonylmethylene)triphenylphosphorane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl 2-(triphenylphosphoranylidene)acetate

L15144-06 L15144-14
CAAAAL15144-03EA 69.9 CAD
CAAAAL15144-03 CAAAAL15144-06 CAAAAL15144-14
(tert-Butoxycarbonylmethylene)triphenylphosphorane 98%
(tert-Butoxycarbonylmethylene)triphenylphosphorane

Purity: 98% CAS No.: 35000-38-5 Molecular Formula: C24H25O2P Formula Weight: 376.44 Melting Point: 146-150° Beilstein Reference: 2759875 UN No.: 2811 MDL No.: MFCD00075545

Formula: C₂₄H₂₅O₂P
MW: 376.436 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00075545
CAS Number: 35000-38-5
EINECS: 412-880-7
UN: 3464
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR